Learn More
Sino Biological Human COQ7 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1219515
Encompass_Preferred
The Human COQ7 ELISA Pair Set is for the quantitative determination of Human COQ7.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human COQ7 coated on a 96-well plate. Standards and samples are added to the wells, and any Human COQ7 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human COQ7 polyclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human COQ7 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86378