Learn More
STEMCELL Technologies Human Cord Blood CD19+ B Cells, Frozen, Size: 1 x 10^6 cells
SDP
Supplier:  STEMCELL Technologies 70013
Encompass
Primary human CD19+ B cells were isolated from umbilical cord blood (CB) mononuclear cells (MNCs) using positive immunomagnetic separation techniques. CB was collected using citrate-phosphate-dextrose (CPD) as an anticoagulant. CD19 is primarily expressed on B cells.Cells were collected using consent forms and protocols approved by either an Institutional Review Board, the Food and Drug Administration, the U.S. Department of Health and Human Services, and/or an equivalent regulatory authority.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.
Catalog No. 50-197-5781