Learn More
STEMCELL Technologies Human Cord Blood CD34+ Cells, Frozen, Size: 1 x 10^6 cells
SDP
Supplier:  STEMCELL Technologies 70008
Encompass
Start experiments confidently with ready-to-use, ethically sourced, human CD34+ stem and progenitor cells. With personalized service, custom products, flexible delivery times, and the option to reserve entire lots to prescreen cells for applications, we help you get the cells you need. Isolated immunomagnetically from umbilical cord blood using positive selection and cryopreserved in serum-free medium, cells are collected using Institutional Review Board (IRB) or other regulatory authority-approved consent forms and protocols. Additional documentation to meet institutional requirements and high-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. CPD is added during collection as an anticoagulant. Contact techsupport@stemcell.com for donor specifications (BMI category, smoking status, ethnicity, etc.). Donors are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C. Certain products are only available in select territories.
Catalog No. NC1074233