Learn More
Biomatik Corporation Human Corin (CRN) ELISA Kit
Supplier:  Biomatik Corporation EKU03456
Encompass_Preferred
Human Corin (CRN) ELISA Kit for detecting CRN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y5Q5. Gene ID: 10699. Target Synonyms: ATC2; CRN; Lrp4; TMPRSS10; Atrial Natriuretic Peptide-Converting Enzyme, Serine Protease; Heart-specific serine proteinase ATC2; Pro-ANP-converting enzyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4557