Learn More
Sino Biological Human coronavirus (HCoV-229E) Nucleocapsid Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40640T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human coronavirus (HCoV-229E) Nucleocapsid ( Catalog#40640-V07E; NP_073556.1; Met1-Asn389). Coronavirus Nucleocapsid specific IgG was purified by Human coronavirus (HCoV-229E) Nucleocapsid affinity chromatography.
Catalog No. 50-224-7385