Learn More
BIOHIPPO Human CTSA Microsample
SDP
Supplier:  BIOHIPPO BHE15211492
Encompass
Human CTSA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CTSA in serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 25 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CTSA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CTSA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CTSA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14835