Learn More
Sino Biological Anti-Cathepsin A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10482RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CTSA (rh CTSA; Catalog#10482-H08H; NP_001121167.1; Met 1-Tyr 480). CTSA specific IgG was purified by human CTSA affinity chromatography.
Catalog No. 50-241-0425