Learn More
BIORTUS USA INC Human CUL4A/RBX1 Protein, no tag, 100UG
Supplier:  BIORTUS USA INC BP1967500A
Encompass_Preferred
Gene Name: CUL4A/RBX1. Uniprot: Q13619/P62877. Construct: Cul4a(A2-A759 end)/RBX1(M1-H108 end). Expression Host: Insect cell. Purity: 90%.
Catalog No. NC2953334