Learn More
Sino Biological Human CXCL5 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1088915
Encompass_Preferred
The Human CXCL5 ELISA Pair Set is for the quantitative determination of Human CXCL5.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CXCL5 coated on a 96-well plate. Standards and samples are added to the wells, and any Human CXCL5 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CXCL5 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CXCL5 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85925