Learn More
Sino Biological Recombinant Human CXCR3 Protein (VLP, Full-length, Flag & His-Strep Tag)
Supplier:  Sino Biological 12979H82H20
Encompass_Preferred
A DNA sequence encoding the Human CXCR3 (P49682) (Met1-Leu368) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.
Catalog No. 50-299-3472