Learn More
Discovery Life Sciences HUMAN CYP2D6 1(VAL374)+REDUCTA
SDP
Supplier:  Discovery Life Sciences 456217
Encompass
Human CYP2D61 + P450Reductase SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP2D61 which is expressed from human CYP2D61 cDNA using a baculovirus expression system
Catalog No. NC9418162