Learn More
Meso Scale Discovery V-PLEX Human Cytokine 30-Plex Kit, multiplex immunoassay, 1 plate, human biomarker measurement
SDP
Supplier:  Meso Scale Discovery K15054D1
Encompass
The MESO SCALE DISCOVERY V-PLEX Human Cytokine 30-Plex Kit is a high-performance multiplex immunoassay designed for the quantitative measurement of 30 human biomarkers associated with inflammatory responses and immune system regulation. This kit ensures accurate and reproducible results, making it suitable for various sample types like serum and plasma. It is analytically validated, offering sensitive detection limits in the sub- to low pg/mL range.
  • High-performance multiplex immunoassay
  • Quantitative measurement of 30 human biomarkers
  • Suitable for cell culture supernatants, serum, plasma, urine
  • Detection limits in the sub- to low pg/mL range
  • Lot-to-lot consistency guaranteed
  • Intra-run CV typically <5%
  • Available for research use only
  • Ships in 1-3 days for 1 plate size
Catalog No. NC2468794
Only null left
Add to Cart
Request a Quote