Learn More
Raybiotech Inc HUMAN CYTOKINE ARRAY C1000 (2)
Supplier:  Raybiotech Inc AAHCYT10002
Encompass_Preferred
RayBio C-Series Human Cytokine Antibody Array 1000 Kit. A combination of Human Cytokine Antibody Array C6 & C7. Detects 120 Human Cytokines. Suitable for all liquid sample types.
Catalog No. 50-104-3083