Learn More
Raybiotech Inc C-Series Human Cytokine Antibody Array 3 Kit.
Supplier:  Raybiotech Inc AAHCYT38
Encompass_Preferred
Detects 42 Humans Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: ENA-78 (CXCL5), GCSF, GM-CSF, GRO alpha/beta/gamma, GRO alpha (CXCL1), I-309 (TCA-3/CCL1), IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-10, IL-12 p40/p70, IL-13, IL-15, IFN-gamma, MCP-1 (CCL2), MCP-2 (CCL8), MCP-3 (MARC/CCL7), M-CSF, MDC (CCL22), MIG (CXCL9), MIP-1 delta (CCL15), RANTES (CCL5), SCF, SDF-1 alpha (CXCL12 alpha), TARC (CCL17), TGF beta 1, TNF alpha, TNF beta (TNFSF1B), EGF, IGF-1, Angiogenin, Oncostatin M, Thrombopoietin (TPO), VEGF-A, PDGF-BB, Leptin
Catalog No. 50-194-8549