Learn More
Raybiotech Inc G-Series Human Cytokine Array 7 Kit.
Supplier:  Raybiotech Inc GSHCYT71
Encompass_Preferred
Semi-quantitative measurement of 40 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: ACE-2, aFGF (FGF-1), Albumin, AMICA, Angiopoietin-4, BAFF, CA19-9, CD163, Clusterin, CRTAM, CXCL14 (BRAK), Cystatin C, Decorin, Dkk-3, DLL1, Fetuin A, FOLR1, Furin, GASP-1 (WFIKKNRP), GASP-2 (WFIKKN), GCSF R (CD114), HAI-2, IL-17 RB, IL-27, LAG-3, LDL R, Pepsinogen 1, RANK (TNFRSF11A), RBP4, SOST, Syndecan-1, TACI (TNFRSF13B), TFPI, Thrombospondin 1, TRAIL R2 (TNFRSF10B/DR5), TRANCE (TNFSF11), Troponin I, uPA, VE-Cadherin (CDH5), WISP-1 (CCN4)
Catalog No. 50-195-0390