Learn More
Raybiotech Inc Quantibody Human Cytokine Array 9 Kit.
Supplier:  Raybiotech Inc QAHCYT92
Encompass_Preferred
Detects 40 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. View protein list on RayBiotech’s website – QAH-CYT-9-2
Catalog No. 50-195-0828