Learn More
Sino Biological Anti-ZIP Kinase/DAPK3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100807T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ZIP Kinase / DAPK3, and purified by antigen affinity chromatography.
Catalog No. 50-240-8581