Learn More
Sino Biological Recombinant Human DDR1 Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Supplier:  Sino Biological 10730H49HB100
Encompass_Preferred
A DNA sequence encoding the Human DDR1 (NP_001189452.2) (Met1-Thr416) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2690