Learn More
American Research Products Inc Human Dehydroepiandrosterone
Supplier:  American Research Products Inc CSBE08226H
Encompass
Human Dehydroepiandrosterone DHEA ELISA Kit from CUSABIO Detection Range 0 1 ng/mL-20 ng/mL Sensitivity 0 05 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26687