Learn More
Sciencell Research Laboratories Human Dermal Fibroblasts-adult, 1 ml volume, > 5 x 10^5 cells/vial, spindle morphology, passage one cryopreserved, negative for HIV-1, HBV, HCV, for research use only
Supplier:  Sciencell Research Laboratories 2320
Encompass
Human Dermal Fibroblasts-adult, 1 ml volume, > 5 x 10^5 cells/vial, spindle morphology, passage one cryopreserved, negative for HIV-1, HBV, HCV, for research use only
Catalog No. NC0757604