Learn More
Sino Biological Human DLL4 ELISA Pair Set 5PP (Sino Biological)
Supplier:  Sino Biological SEK101715
Encompass_Preferred
Each vial contains 170 ng of recombinant DLL4. Reconstitute standard powder with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in dilution buffer, and a high standard of 4000 pg/mL is recommended.
Catalog No. 50-237-0643