Learn More
CHIMERX Human DNA Polymerase Alpha, 200 u
Supplier:  CHIMERX 1075200U
Encompass
Exclusively from CHIMERx, Human DNA Polymerase Alpha is a replication enzyme used to test the toxicity of drugs on key human replication systems.DescriptionThe simplest DNA Polymerase known in both size and catalysisReplication enzyme involved in initiation of DNA replication in eukaryotesA multi-subunit complex, which inhibits DNA polymerase and primase activitiesIsolated from HeLa cells by immunoaffinity chromatography (1)ApplicationsUsed to test in vitro replication capabilities along with other enzymesUsed to test the toxicity of drugs on key human replication systemsUsed in DNA replication experimentsUnit DefinitionOne unit is the amount of enzyme required to incorporate 1 nmole of total nucleotide into acid-insoluble form in 60 minutes at 37°C
Catalog No. NC2751899