Learn More
CHIMERX Human DNA Polymerase Gamma
Supplier:  CHIMERX 1076200U
Encompass
Exclusively from CHIMERx, Human DNA Polymerase Gamma is a mitochondrial enzyme used for testing the toxicity of drugs on critical human oxidative functions.DescriptionMitochondrial enzyme used for testing the toxicity of drugs on critical human oxidative functions and testing replication of mitochondrial DNAUnit DefinitionOne unit is the amount of enzyme required to incorporate 1 pmole of total nucleotide into acid-insoluble form in 60 minutes at 37°C
Catalog No. NC2751905