Learn More
American Research Products Inc Human Ectonucleotide pyrophosp
Supplier:  American Research Products Inc CSBEL007680HU
Encompass
Human Ectonucleotide pyrophosphatase/phosphodiesterase family member 2 (ENPP2) ELISA kit from CUSABIO Detection Range 25 ng/mL-2000 ng/mL Sensitivity 31 25 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27920