Learn More
Raybiotech Inc Human eNOS/NOS3 ELISA Kit
Supplier:  Raybiotech Inc ELHENOS1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.4 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human eNOS.
Catalog No. 50-194-9219