Learn More
American Research Products Inc Human eosinophil cationic prot
Supplier:  American Research Products Inc CSBE11729H
Encompass
Human eosinophil cationic protein ECP ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26844