Learn More
Sino Biological Human EpCAM/TROP1 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1069415
Encompass_Preferred
The Human EpCAM/TROP-1/TACSTD1 ELISA Pair Set is for the quantitative determination of Human EpCAM/TROP-1/TACSTD1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human EpCAM/TROP-1/TACSTD1 coated on a 96-well plate. Standards and samples are added to the wells, and any Human EpCAM/TROP-1/TACSTD1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human EpCAM/TROP-1/TACSTD1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human EpCAM/TROP-1/TACSTD1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86373