Learn More
ACROBiosystems ACROBiosystems Human Ephrin-A3 / EFNA3 Protein, Fc Tag
Supplier:  ACROBiosystems EA3H5258200UG
Encompass_Preferred
Recombinant Protein;200UG;Human Ephrin-A3, Fc Tag (EA3-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 213 (Accession # AAI10407).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48 kDa. The protein migrates as 57-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6626