Learn More
Abcam Human Exodus 2 ELISA Kit (CCL21), 1 x 96 Tests
Supplier:  Abcam AB1937591X96TESTS
Encompass_Preferred
Human Exodus 2 ELISA Kit (CCL21) is a single-wash 90-min Simplestep used to quantify Human Exodus 2 (CCL21) with a sensitivity of 3.8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2582839