Learn More
BIOHIPPO Human F-actin Microsample
SDP
Supplier:  BIOHIPPO BHE15212428
Encompass
Human F-actin Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of F-actin in Serum plasma tissue homogenates It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 18 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human F-actin Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human F-actin Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human F-actin biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14968