Learn More
Abcam Human Factor IX ELISA Kit (with plasma controls), 1 x 96 Tests
Supplier:  Abcam AB1685461X96TESTS
Encompass_Preferred
Human Factor IX ELISA Kit (with plasma controls) is a Sandwich (quantitative) ELISA kit for the measurement of Human Factor IX (with plasma controls) in Human in Cerebral Spinal Fluid, Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1983577