Learn More
Haematologic Technologies Inc HUMAN FACTOR VA 50UG
Supplier:  Haematologic Technologies Inc HCVA0110
Encompass_Preferred
Coagulation protein, Human Factor Va, 50 micrograms, Activated from of factor V, Mode of action: cofactor for factor Xa in the prothrombinase complex, Research use only, 50% (v/v) glycerol/water + 2mM CaCl2, store at -20 Celsius.
Catalog No. 50-883-310