Learn More
Haematologic Technologies Inc HUMAN FACTOR X 100UG
Supplier:  Haematologic Technologies Inc HCX0050
Encompass_Preferred
Coagulation Protein, Human Factor X, 100 micrograms, Purifed from plasma, Zymogen, MW=58,900, Structure: two subunits, Mr=16,200 and 42,000, NH2-terminal gla domain, and two EGF domains, Research use only, 50% (v/v) glycerol/water, Store at -20 Celsius.
Catalog No. 50-883-355