Learn More
American Research Products Inc Human Fc gamma RI / CD64 protein 1MG
Supplier:  American Research Products Inc HUGR1U
Encompass
The sequence of the extracellular domain of human CD64 (Gln 16-Leu 281) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.
Catalog No. NC3255299