Learn More
American Research Products Inc Human Fc region of immunoglobu
Supplier:  American Research Products Inc CSBE11223H
Encompass
Human Fc region of immunoglobulin G Fcy ELISA Kit from CUSABIO Detection Range 0 027 g/mL-20 g/mL Sensitivity 0 024 g/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29068