Learn More
BIOHIPPO Human FcgBP Microsample
SDP
Supplier:  BIOHIPPO BHE15211037
Encompass
Human FcgBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FcgBP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 114 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FcgBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FcgBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FcgBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14468