Learn More
ACROBiosystems Human FcRn binding Kit (TR-FRET) 500 Tests
Supplier:  ACROBiosystems FRT01500TESTS
Encompass_Preferred
The Human FcRn binding Kit (TR-FRET) is based on a homogeneous (no wash) competition TR-FRET technology (Time-Resolved Fluorescence Resonance Energy Transfer) to measure the interaction between human FcRn and antibody drug candidates or FcRn inhibitors. It is designed to facilitate the half-life evaluation of antibody drug candidates, high-throughput screening of FcRn inhibitors within 0.5-1 hours. It can also be used as a universal detection tool to identify the ability of antibody drugs to bind to FcRn.Please note that the TR-FRET Sample Dilution Buffer and Detection Buffer are formulated for distinct purposes and should not be mixed or interchanged, as improper use may lead to unexpected or inconsistent experimental results. The Sample Dilution Buffer is intended solely for sample dilution, whereas the Detection Buffer is specifically designed for diluting Donor and Acceptor reagents.For experiments performed under different pH c
Catalog No. NC3987734