Learn More
Abcam Human Ferritin Light Chain ELISA Kit- Intracellular, 1 x 96 Tests
Supplier:  Abcam AB3168941X96TESTS
Encompass_Preferred
Human Ferritin Light Chain ELISA Kit- Intracellular is a single-wash 90-min Simplestep used to quantify Human Ferritin Light Chain with a sensitivity of 8.589 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-1642