Learn More
Abcam Human Galectin 1 Matched Antibody Pair Kit, 5 x 96 Tests
Supplier:  Abcam AB2181705X96TESTS
Encompass_Preferred
Human Galectin 1 Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Human Galectin 1 by sandwich ELISA method.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-7905