Learn More
Bachem Ghrelin (human) Trifluoroacetate
SDP
Supplier:  Bachem 4033077.1000
Encompass_Preferred
Ghrelin (human) Trifluoroacetate, H-Gly-Ser-Ser(octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH, 258279-04-8, C149H249N47O42, Mr 3370.91, 1mg. Human ghrelin (GHR), the endogenous ligand of the growth hormone secretagogue receptor (GHS-R), potently stimulates GH release. The Ser3 octanoyl ester turned out to be essential for generating this activity. Ghrelin plays an important role in the short-time regulation of appetite and in the long-time regulation of energy balance and glucose homeostasis. A number of studies showed that ghrelin is also involved in the regulation of gastrointestinal, cardiovascular, and immune functions as well as in bone metabolism.
Catalog No. 50-194-6890