Learn More
FUJIFILM BIOSCIENCES INC Human Glioma Culture Media
Supplier:  FUJIFILM BIOSCIENCES INC PB50019
Encompass_Preferred
Cytokines for cell culture of human DIPG and other glioma cells. A glioma is a generic term used to describe tumors that arise from the glia tissue of the brain that supports and surrounds neurons. Diffuse Intrinsic Pontine Glioma (DIPG) is a pediatric brain tumor that originates in the brainstem. Cells isolated from these childhood tumors are successfully cultured ex vivo with EGF, FGF-basic (or FGF-2), and PDGFs (AA and BB).
Catalog No. 50-289-2145