Learn More
Biomatik Corporation Human Glutathione S Transferase Mu 1 (GSTM1) ELISA Kit
Supplier:  Biomatik Corporation EKU04494
Encompass_Preferred
Human Glutathione S Transferase Mu 1 (GSTM1) ELISA Kit for detecting GSTM1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.16ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09488. Gene ID: 2944. Target Synonyms: GST-M1; GST1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GTH4; GTM1; H-B; MU1; GST HB subunit 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4717