Learn More
ACROBiosystems PE-Labeled Human Glypican 3 / GPC3 Protein, His TagStar Staining
Supplier:  ACROBiosystems GP3HP2H925TESTS
Encompass_Preferred
PE-Labeled Human Glypican 3 Protein, His Tag (GP3-HP2H9) is produced via conjugation of PE to Human Glypican 3 Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Glypican 3 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).
Catalog No. 50-233-1049