Learn More
ILEX LIFE SCIENCES LLC Human Gonadotropin-Releasing Hormone (GnRH), Synthetic Protein, 10 mg
SDP
Supplier:  ILEX LIFE SCIENCES LLC A2261010
Encompass_Preferred
Gonadotropin-Releasing Hormone (GnRH) Human, Synthetic also known as Luteinizing-hormone releasing hormone (LHRH), is a single, non-glycosylated, polypeptide chain containing 10 amino acids and has a 1182.2 Dalton molecular mass. GnRH is a peptide hormone responsible for the release of FSH and LH from the anterior pituitary. MOLECULAR FORMULA: C55H75N17O13.C2H4O2. CAS NO.: 71447-49-9. AMINO ACID SEQUENCE: Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2. PURITY: Greater than 98.0%. FORMULATION: Sterile-filtered white lyophilized (freeze-dried) powder. The synthetic GnRH was lyophilized from a concentrated (1 mg/ml) solution with no additives. OTHER NAMES: Luteinizing hormone-releasing hormone I, Luteinising-hormone releasing hormone, LHRH, GRH, GNRH, LNRH, LH-RH I, Gonadotropin-releasing hormone I, GnRH-I, GNRH1, Luliberin I, Gonadorelin, Gonadoliberin I, Gonadoliberin-1, Progonadoliberin I, Progonadoliberin-1, GnRH-associated peptide 1, GnRH-associated peptide I.
Catalog No. 50-233-9512