Learn More
American Research Products Inc Human Group IIF secretory phos
Supplier:  American Research Products Inc CSBEL018095HU
Encompass
Human Group IIF secretory phospholipase A2 (PLA2G2F) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28214