Learn More
BIORTUS USA INC Human HCK Protein, 100UG
Supplier:  BIORTUS USA INC BP0582901A
Encompass_Preferred
Gene Name: HCK. Uniprot: P08631. Construct: 8His-TEV-HCK(I81-P526 end)(Q523E, Q524E, Q525I). Expression Host: Insect cell. Purity: 90%.
Catalog No. NC2953347