Learn More
Ethos Biosciences hemoglobin HbA0 standard blood 1/EA
Supplier: Ethos Biosciences HBA0
Hemoglobin A0 is prepared from lysed human blood cells by sequential ion-exchange It is a mixture of non-glycated and glycated hemoglobin A0 fractions but is essentially free of HbA1a HbA1b HbA1c and HbF The protein is validated by HPIEC methods and is suitable as a primary standard in chromatographic or immunologic analyses The reported concentration is determined by Drabkins assay The material is supplied in liquid form in dilute potassium cyanide-phosphate buffer The concentration varies with manufacturing lot but the protein may be used directly by dilution in buffer in most experimental methods It is shipped frozen at -20oCShort term storage should be at 4oC Long term storage should be at -60oC Repeated cycles of freezing and thawing should be avoided