Learn More
Bioworld Human Heparanase-1 (Recombinant), 100 ng
Supplier:  Bioworld 220603521
Encompass_Preferred
Human Heparanase-1, Recombinant (HPA1)Heparanase is an endo β-D-glucouronidase, which degrades heparan sulfate side chains of heparan sulfate proteoglycans (HSPGs) in the extracellular matrix. Recombinant Heparanase protein HPA1 is produced in CHO cells.Application: Positive control for western blot analysis
Catalog No. 50-253-449