Learn More
ACROBiosystems Alexa Fluor 647-Labeled Human Her2 / ErbB2 Protein, His TagStar Staining
Supplier: ACROBiosystems HE2HA2H525UG
Alexa Fluor 647-Labeled Human Her2, His Tag (HE2-HA2H5) is produced via conjugation of AF647 to Human Her2, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Her2, His Tag is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1).