Learn More
ATCC Human herpesvirus 5; HCMV Strain AD-169; Quantitative DNA
Supplier:  ATCC VR538DQ
Encompass
Quantitative genomic DNA from Human herpesvirus 5 strain AD-169 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load and for other molecular biology applications.
Catalog No. 50-238-3420
 Restricted Item